1,2,4,6-Tetra-O-acetyl-3-O-allyl-β-D-glucopyranose ≥98.0% (by HPLC)
Catalog # TCIAT2449-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:117.0 to 121.0 deg-C
- Specific rotation [a]20/D:-1.0 to -4.0 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:388.37 g/mol
Storage Temperature:Freezer
CAS Number:39698-00-5
Formula:C₁₇H₂₄O₁₀