1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-ɑ-D-mannopyranose ≥98.0% (by HPLC)
Catalog # TCIAT1733-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+75.0 to +85.0 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:373.32 g/mol
Storage Temperature:Refrigerator
CAS Number:68733-20-0
Formula:C₁₄H₁₉N₃O₉
MDL Number:MFCD01321201