Order Entry
Sweden
ContactUsLinkComponent
 
cis-15-Tetracosenoic acid ≥95.0% (by GC)
  TCIAT1642-1G
 :  TCI
 :  
cis-15-Tetracosenoic acid ≥95.0% (by GC)
  TCIAT1642-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    42.0 to 46.0 deg-C

 

 

 :
 :366.63 g/mol
 :GHS07
 :Refrigerator
 :506-37-6
 :C₂₄H₄₆O₂
 :MFCD00010507