Order Entry
Sweden
ContactUsLinkComponent
 
Octadecyl-p-toluenesulphonate ≥98.0% (by GC)
Catalog # TCIAT1471-5G
Supplier:  TCI
CAS Number:  
Octadecyl-p-toluenesulphonate ≥98.0% (by GC)
Catalog # TCIAT1471-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    56.0 to 59.0 deg-C

Specification Test Results

About this item

 :
 :424.68 g/mol
CAS Number:3386-32-1
Formula:C₂₅H₄₄O₃S
MDL Number:MFCD00191588