Skip to Header
Skip to Main Content
Skip to Footer
Avancerad
Avancerad
SDS
Certifikat
Tillverkardata
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromoisatin
7-Bromoisatin ≥97%
7-Bromoisatin ≥97%
Irritant
7-Bromoisatin ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
7-Bromoisatin ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
20780-74-9
:
Formula:
C₈H₄BrNO₂
MDL Number:
MFCD00774354
Melting Pt:
195…199 °C
:
226.03 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH32843
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation