Nɑ-Tosyl-L-arginine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:1784-03-8
:
Formel:C₁₄H₂₂N₄O₄S·HCl
MDL-nummer:MFCD00012578
:378.87 g/mol
EINECS:217-235-1
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:146.0 to 150.0 deg-C
- Specific rotation [a]20/D:-13.0 to -15.0 deg (C=4, H2O)
- Solubility in Methanol:almost transparency