(Toluene-3,4-dithiolato)zinc(II) ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:29726-21-4
:
Formula:C₇H₈S₂
MDL Number:MFCD00005863
:219.63 g/mol
EINECS:249-812-9
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together