Spiro[fluorene-9,9'-xanthene]-3',6'-diol ≥98.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:4081-00-9
:
Formel:C₂₅H₁₆O₃
:364.40 g/mol
Specifikationstestresultat
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %