Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:5619-04-5
:
Formel:C₄H₁₀ClNO₃
MDL-nummer:MFCD00012593
:155.578 g/mol
EINECS:227-047-1
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %