Order Entry
Sweden
ContactUsLinkComponent
 
L-Serine benzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
  TCIAS0550-5G
 :  TCI
 :  
undefined
L-Serine benzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
  TCIAS0550-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -10.0 to -13.0 deg (C=1, H2O)

 

 

 :
 :231.6 g/mol
 :60022-62-0
 :C₁₀H₁₃NO₃·HCl
 :MFCD00038955