

Sodium hydride 60% dispersion in paraffin liquid
Supplier: TCI
Certificates
About this item
CAS Number:7646-69-7
:
Formula:HNa
MDL Number:MFCD00003471
Merck Index:14,08625
ADR:4.3,I
:24 g/mol
EINECS:231-587-3
UN:1427
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Gray to Dark blue powder to crystal
- Concentration of NaH:57.0 to 63.0 w/w%