Order Entry
Sweden
ContactUsLinkComponent
SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0% (by GC) cross-linking reagent
Irritant
SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0% (by GC) cross-linking reagent
Supplier:  TCI
Certificates
Irritant
SATA (N-Succinimidyl S-Acetylthioglycolate) ≥94.0% (by GC) cross-linking reagent
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 94.0 %
  • Melting point:
    95.0 to 101.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together