Synephrine tartrate ≥98.0% (by HPLC, titration analysis)
TCIAS0233-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:182.0 to 186.0 deg-C
:
:484.50 g/mol
:240-647-8
:16589-24-5
:C₉H₁₃NO₂
:MFCD00151120