


SILREST column conditioner for gas chromatography
Catalog # TCIAS0162-12ML
Supplier: TCI
CAS Number:
Specifications
- Pk:12 mL
- Seq:2288
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- High boiling impurity of Silylation agent:No impurity higher than Isopropyl myristate (50 ppm) after 5 min + hold up time
Specification Test Results
About this item
:
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
MDL Number:MFCD00148405