Order Entry
Sweden
ContactUsLinkComponent
Ruthenium(II) nitrosyl chloride ≥97.0% (by titrimetric analysis)
Irritant
Ruthenium(II) nitrosyl chloride ≥97.0% (by titrimetric analysis)
Catalog # TCIAR0086-1G
Supplier:  TCI
CAS Number:  
Irritant
Ruthenium(II) nitrosyl chloride ≥97.0% (by titrimetric analysis)
Catalog # TCIAR0086-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Yellow to Amber to Dark red powder to crystal
  • Purity (Argentmetric Titration):
    min. 97.0 % (calcd.on anh.substance)
  • Water:
    6.0 to 9.0 %

Specification Test Results

About this item

 :
 :237.43 g/mol
EINECS:242-651-5
 :GHS07
CAS Number:18902-42-6
Formula:Ru(NO)Cl₃
MDL Number:MFCD00049765