Order Entry
Sweden
ContactUsLinkComponent
Rose Bengal
Rose Bengal
Catalog # TCIAR0041-25G
Supplier:  TCI
CAS Number:  
Rose Bengal
Catalog # TCIAR0041-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Dark red to Brown powder to crystal
  • Lambda max.:
    547.0 to 551.0 nm (0.02mol/L AcONH4 sol.)
  • Absorbance (E1%1cm):
    min. 900 (0.02mol/L AcONH4 sol., 547.0 to 551.0 nm)

Specification Test Results

About this item

 :
 :1017.63 g/mol
EINECS:211-183-3
CAS Number:632-69-9
 :MFCD00005043
Formula:C₂₀H₂Cl₄I₄O₅Na₂
MDL Number:MFCD00151169