Phenylbis(phenylthio)methane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7695-69-4
:
Formula:C₁₉H₁₆S₂
MDL Number:MFCD00092116
:308.46 g/mol
Boiling Pt:184°C
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:48.0 to 52.0 deg-C