Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside ≥98.0% (by HPLC)
TCIAP2521-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
:
:440.46 g/mol
:85°C
:Refrigerator
:108032-93-5
:C₂₀H₂₄O₉S
:MFCD01862644