Order Entry
Sweden
ContactUsLinkComponent
3-Phosphonobenzoic acid ≥98.0% (by HPLC, titration analysis)
Corrosive
3-Phosphonobenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP2458-200MG
Supplier:  TCI
CAS Number:  
undefined
Corrosive
3-Phosphonobenzoic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP2458-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :202.10 g/mol
UN:3261
ADR:8,II
 :GHS05
CAS Number:14899-31-1
Formula:C₇H₇O₅P
MDL Number:MFCD00159422