Phenolphthalein Diphosphate Pentasodium salt hydrate ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:2382-56-1
:
Formula:C₂₀H₁₃Na₅O₁₁P₂·xH₂O
:606.21 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 95.0 area%
- Water:max. 30.0 %
- Ethanol (NMR):max. 10.0 %