
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran ≥97.0% (by titrimetric analysis)
TCIAP2004-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Amber to Brown powder to crystal
- Purity (Argentmetric Titration):min. 97.0 %
:
:923.09 g/mol
:480-140-0
:GHS07
:250220-36-1
:C₅₁H₇₆Cl₂P₂Ru
:MFCD07369034