Phenyl-3,4,6-tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-β-D-glucopyranoside ≥98.0% (by HPLC)
Catalog # TCIAP1866-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:572.83 g/mol
Melting Pt:143°C
Storage Temperature:Freezer
CAS Number:187022-49-7
:MFCD15072189
Formula:C₂₁H₂₄Cl₃NO₉S
MDL Number:MFCD11112181