
(R)-Piperidine-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1723-00-8
:
Formula:C₆H₁₁NO₂
MDL Number:MFCD00064346
:129.16 g/mol
Melting Pt:270°C
EINECS:217-024-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+24.0 to +29.0 deg (C=1, H2O)