Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Palladium
Palladium 5% on barium carbonate
Palladium 5% on barium carbonate
Palladium 5% on barium carbonate
TCIAP1701-10G
:
TCI
:
Palladium 5% on barium carbonate
TCIAP1701-10G
:
TCI
:
Pk:
10 g
Seq:
3236
Content (Palladium):
4.50 to 5.50 %
Water:
max. 3.0 %
:
:
106.420 g/mol
:
14,06989
:
231-115-6
:
1564
:
6.1,III
:
GHS06
:
GHS08
:
7440-05-3
:
Pd
:
MFCD03427451