Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside ≥98.0% (by HPLC)
: TCI
:24404-53-3
:
:C₂₀H₂₄O₉S
:MFCD06657849
:440.46 g/mol
:Freezer
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+4.0 to +6.0 deg (C=1, CHCl3)