



2,4,6-Trinitrobenzenesulphonic acid (TNBS) ∼1% in DMF for detection of primary amines
Supplier: TCI
Certificates
About this item
CAS Number:2508-19-2
:
Formula:C₆H₃N₃O₉S
MDL Number:MFCD00064382
:293.17 g/mol
Flash Pt:58°C
EINECS:219-717-7
UN:2265
ADR:3,III
:GHS02
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:Light yellow to Yellow clear liquid
- Concentration (Neutralization titration):0.9 to 1.1 w/w%