Order Entry
Sweden
ContactUsLinkComponent
Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0% (by GC)
Irritant
Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    185.0 to 189.0 deg-C
  • Solubility in Toluene:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together