(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Katalog # TCIAP1278-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:+36.0 to +39.0 deg (C=2, EtOH)
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:215.68 g/mol
EINECS:231-383-4
CAS Number:7524-50-7
Formula:C₁₀H₁₄ClNO₂
MDL Number:MFCD00012489