Pentaethylene glycol monomethyl ether ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23778-52-1
:
Formula:C₁₁H₂₄O₆
MDL Number:MFCD00045995
:252.31 g/mol
Storage Temperature:Refrigerator
EINECS:245-874-6
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %