Polyvinyl alcohol n ∼2,000, (degree of saponification ca. 80mol%)
: TCI
:9002-89-5
:25213-24-5
:[-CH₂CH(OH)-]n
:MFCD00081922
:14,07585
- Appearance:White to Orange to Green powder to crystal
- Solubility in Water:almost transparency
- degree of saponification:78.0 to 82.0 mol%
- Suitability for Absorptiometry:to pass test