Order Entry
Sweden
ContactUsLinkComponent
 
Pentaerythritol tetrastearate ≥95.0% (by titrimetric analysis) (contains palmitate)
 :  TCI
 
Pentaerythritol tetrastearate ≥95.0% (by titrimetric analysis) (contains palmitate)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ester Value):
    min. 95.0 %
  • Melting point:
    62.0 to 66.0 deg-C