Order Entry
Sweden
ContactUsLinkComponent
(±)-1,2-Propanediol ≥99.0% (by GC)
(±)-1,2-Propanediol ≥99.0% (by GC)
Katalog # TCIAP0485-500G
Leverantör:  TCI
CAS-nummer:  
undefined
(±)-1,2-Propanediol ≥99.0% (by GC)
Katalog # TCIAP0485-500G
Leverantör:  TCI
CAS-nummer:  

Specifikationer

  • Pk:
    500 g
  • Seq:
    3645

Specifikationer

Specifikationstestresultat

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specifikationstestresultat

Om denna produkt

 :
 :76.1 g/mol
Merck Index:14,07855
Kokande Pt:188°C
Smält Pt:-59°C
Smält Pt:–59°C
Flash Pt:107°C
EINECS:200-338-0
 :GHS07
CAS-nummer:57-55-6
Formel:CH₃CH(OH)CH₂OH
MDL-nummer:MFCD00064272