Polyvinyl alcohol n=1750±50
Supplier: TCI
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922
Merck Index:14,07585
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Solubility in hot Water:almost transparency
- Drying loss:max. 7.0 %
- degree of saponification:97.0 to 100.0 mol%