
Sodium Pyridine-4-sulphinate ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:116008-37-8
:
Formula:C₅H₄NNaO₂S
MDL Number:MFCD30502924
:165.14 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %