Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane ≥95.0% (by GC)
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane ≥95.0% (by GC)
:
TCI
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane ≥95.0% (by GC)
:
TCI
:
16066-09-4
:
:
C₈H₂₆O₃Si₄
:
MFCD00053664
:
282.63 g/mol
:
177°C
:
63°C
:
240-209-6
:
GHS07
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 95.0 %
Manual Bioscience and Analyticalscience