4-Nitro-DL-phenylalanine hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:207569-25-3
:
Formula:C₉H₁₀N₂O₄·xH₂O
MDL Number:MFCD00007384
:210.19 g/mol (anhy)
EINECS:220-868-6
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Water:max. 10.0 %