Order Entry
Sweden
ContactUsLinkComponent
 
Neryl acetate ≥95.0% (by GC)
  TCIAN0463-25ML
 :  TCI
 :  
Neryl acetate ≥95.0% (by GC)
  TCIAN0463-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :196.29 g/mol
 :140°C
 :107°C
 :205-459-2
 :141-12-8
 :C₁₂H₂₀O₂
 :MFCD00063205