Nerolidol (cis and trans mixture) cis- and trans-mixture ≥97.0% (by GC)
Catalog # TCIAN0454-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 % (total of isomer)
Specification Test Results
About this item
Synonyms: 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, Nerolidol, 3,7,11-Trimethyldodeca-1,6,10-trien-3-ol
:
:222.37 g/mol
Merck Index:14,06476
Boiling Pt:146°C
Flash Pt:135°C
EINECS:230-597-5
:GHS07
CAS Number:7212-44-4
Formula:C₁₅H₂₆O
MDL Number:MFCD00008911