Order Entry
Sweden
ContactUsLinkComponent
 
2-(4-Methoxyphenyl)-1,3-dithiane ≥98.0% (by GC)
 :  TCI
 
2-(4-Methoxyphenyl)-1,3-dithiane ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    117.0 to 121.0 deg-C