Mubritinib ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:366017-09-6
:
Formula:C₂₅H₂₃F₃N₄O₂
MDL Number:MFCD09954135
:468.5 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:158.0 to 162.0 deg-C