4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:36476-29-6
:
Formula:C₁₈H₂₁NO₈
MDL Number:MFCD00067658
:379.365 g/mol
Storage Temperature:Freezer
EINECS:253-052-3
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):3.40 to 4.00 %
- Specific rotation [a]20/D:+23.0 to +26.0 deg (C=0.4, DMSO)
Specification Test Results
Frequently Bought Together