Order Entry
Sweden
ContactUsLinkComponent
(R)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)
Irritant
(R)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
(R)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    123.0 to 127.0 deg-C
  • Specific rotation [a]20/D:
    +1.5 to +3.5 deg (C=1,H2O)

Specification Test Results

Frequently Bought Together