
Methyl 3-Amino-5-bromo-2-methylbenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1000342-11-9
:
Formula:C₉H₁₀BrNO₂
MDL Number:MFCD08690071
:244.09 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %