

Toluene-α-sulphonyl fluoride
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:329-98-6
:
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424
:174.2 g/mol
Boiling Pt:80…85 °C (0.3 mmHg)
Melting Pt:91…92 °C
Storage Temperature:Ambient
EINECS:206-350-2
UN:2923
ADR:8,III
:GHS06
:GHS05