
Methyl pipecolinate hydrochloride ≥98.0% (by titrimetric analysis)
: TCI
:32559-18-5
:MFCD22123553
:C₇H₁₄ClNO₂
:179.64 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %