Order Entry
Sweden
ContactUsLinkComponent
2-Methoxytropone ≥98.0% (by GC)
2-Methoxytropone ≥98.0% (by GC)
Catalog # TCIAM2576-1G
Supplier:  TCI
CAS Number:  
undefined
2-Methoxytropone ≥98.0% (by GC)
Catalog # TCIAM2576-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    130.0 to 134.0 deg-C

Specification Test Results

About this item

 :
 :136.15 g/mol
Boiling Pt:175°C
CAS Number:2161-40-2
Formula:C₈H₈O₂
MDL Number:MFCD01734435