
3-Methyladenine
Supplier: TCI
Certificates
About this item
CAS Number:5142-23-4
:
Formula:C₆H₇N₅
MDL Number:MFCD00010531
:149.16 g/mol
EINECS:225-908-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %