Skip to Header
Skip to Main Content
Skip to Footer
Avancerad
Avancerad
SDS
Certifikat
Tillverkardata
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-3-cyclopropyl-3-oxopropanoate
Methyl-3-cyclopropyl-3-oxopropanoate ≥96.0% (by GC)
Methyl-3-cyclopropyl-3-oxopropanoate ≥96.0% (by GC)
:
TCI
Methyl-3-cyclopropyl-3-oxopropanoate ≥96.0% (by GC)
:
TCI
:
32249-35-7
:
:
C₇H₁₀O₃
:
MFCD00274090
:
142.1500 g/mol
:
80°C
:
74°C
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 96.0 %