Order Entry
Sweden
ContactUsLinkComponent
Metanol-D4
Flammable
Irritant
Health
Metanol-D4
Catalog # TCIAM1869-10ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Metanol-D4
Catalog # TCIAM1869-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Förp.:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Degree of deuteration:
    min. 99.8 atom%D

Specification Test Results

About this item

 :
 :36.01 g/mol
Boiling Pt:66°C
Melting Pt:-98°C
Melting Pt:–98°C
Flash Pt:11°C
EINECS:212-378-6
UN:1230
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637