4-Methoxyphenyl-4-O-acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:140615-77-6
:
Formula:C₃₇H₃₅NO₉
:637.69 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+83.0 to +87.0 deg (C=0.8, CHCl3)
Specification Test Results
Frequently Bought Together